Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA003421

Sigma-Aldrich

Anti-MICAL3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA0819, Anti-microtubule associated monoxygenase, calponin and LIM domain containing 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

YAYDIEDGKFYCKPHYCYRLSGYAQRKRPAVAPLSGKEAKGPLQDGATTDANGRANAVASSTERTPGSGVNGLEEPSIAKRLRGTPERIELENYRLSLRQAEALQEVPEETQAEHNLSSVLDTGAEEDVASRSARRA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MICAL3(57553)

General description

MICAL3 (microtubule associated monooxygenase, calponin and LIM domain containing 3) is a human gene belonging to the MICAL family of flavoprotein monooxygenases. It is located on chromosome 22q11.21.

Immunogen

microtubule associated monoxygenase, calponin and LIM domain containing 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Microtubule-associated monoxygenase, calponin and LIM domain containing 3, also known as MICAL3, possesses the monooxygenase activity required to regulate its own turnover and the simultaneous remodeling of protein complexes that are involved in vesicle-docking and in which it is engaged.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73608.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilya Grigoriev et al.
Current biology : CB, 21(11), 967-974 (2011-05-21)
Rab6 is a conserved small GTPase that localizes to the Golgi apparatus and cytoplasmic vesicles and controls transport and fusion of secretory carriers [1]. Another Rab implicated in trafficking from the trans-Golgi to the plasma membrane is Rab8 [2-5]. Here
Madhushika Ratnayake et al.
BMC medical genetics, 13, 12-12 (2012-03-06)
A genome-wide association scan with subsequent replication study that involved over 67,000 individuals of European ancestry has produced evidence of association of single nucleotide polymorphism rs2277831 to primary osteoarthritis (OA) with a P-value of 2.9 × 10(-5). rs2277831, an A/G

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service