Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA003314

Sigma-Aldrich

Anti-RHOXF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Paired-like homeobox protein PEPP-2, Anti-Rhox homeobox family member 2, Anti-Testis homeobox gene 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:2500-1:5000

immunogen sequence

DQCSQYMTSLLSPAVDDEKELQDMNAMVLSLTEEVKEEEEDAQPEPEQGTAAGEKLKSAGAQGGEEKDGGGEEKDGGGAGVPGHLWEGDLEGTSGSDGNVEDSDQSEKEPGQQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOXF2(84528)

General description

RHOXF2 (Rhox homeobox family, member 2) gene encodes a protein belonging to the RHOX family. It is localized to human chromosome Xq24. The gene contains a Pem DNA homeobox sequence that encodes a 60 amino acid homeodomain protein that associates with DNA. The RHOX family of genes contains two introns and four exons that encode a transcription factor that is specifically expressed in the testis.

Immunogen

Rhox homeobox family member 2 recombinant protein epitope signature tag (PrEST)

application

Anti-RHOXF2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

RHOXF2 (Rhox homeobox family, member 2) gene encodes a 288 amino acid protein that functions as a transcription factor that may have a role in carcinogenesis. Rhox proteins are expressed in germ cells, embryonic cells and somatic cells of reproductive tissue. They may function in embryonic, postnatal and adult development, and in the development of male and female reproductive systems. It may be a candidate gene for spermatogenesis failure.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74383

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christophe Frainais et al.
Basic and clinical andrology, 24, 3-3 (2014-01-01)
Genes involved in testicular differentiation, spermatogenesis, proliferation and apoptosis of germ cells have been shown to evolve rapidly and display rapid DNA changes. These genes are therefore good candidates for explaining impairments in spermatogenesis. Initial studies of some of these
Anthony Arnoldo et al.
Genome medicine, 6(4), 32-32 (2014-06-20)
Target identification is a critical step in the lengthy and expensive process of drug development. Here, we describe a genome-wide screening platform that uses systematic overexpression of pooled human ORFs to understand drug mode-of-action and resistance mechanisms. We first calibrated
Fumi Shibata-Minoshima et al.
International journal of oncology, 40(1), 93-98 (2011-08-30)
Multiple mutations contribute to establish cancers. We have searched for potential oncogenes by screening cDNA libraries derived from gastric cancer cell lines, pancreatic cancer cell lines and glioma cell lines, using retrovirus-mediated expression cloning. Two types of interleukin-3 (IL-3)-dependent cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service