Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA002704

Sigma-Aldrich

Anti-MAPK15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Extracellular signal-regulated kinase 8 antibody produced in rabbit, Anti-Mitogen-activated protein kinase 15 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

KTDAQRTFREITLLQEFGDHPNIISLLDVIRAENDRDIYLVFEFMDTDLNAVIRKGGLLQDVHVRSIFYQLLRATRFLHSGHVVHRDQKPSNVLLDANCTVKLCDF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAPK15(225689)

General description

MAPK15 (mitogen-activated protein kinase 15) is an atypical member of MAPK family with a conserved PCNA-interacting protein (PIP) box and the PXXXP (proline rich) motif in carboxyl terminal end. It exhibits high kinase activity. It is localized at the Golgi complex. It possesses an atypically long C-terminal domain for auto-phosphorylation of its Thr-X-Tyr motif.[1]

Immunogen

Mitogen-activated protein kinase 15 recombinant protein epitope signature tag (PrEST)

Application

Anti-MAPK15 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

MAPK15 (mitogen-activated protein kinase 15) is involved in the proliferating cell nuclear antigen (PCNA) protein stability. It is a major regulator of PCNA complex formation in mammary epithelial cells. In association with homologue of murine double minute (HDM2), it prevents the loss of PCNA during DNA damage and mitotic failure.[1] It plays a role in the DNA repair of single strand breaks through a proteasome-dependent mechanism. In the Golgi complex it inhibits COP-I vesicle formation, which is essential for the formation of transport carriers. It also restrict the export of GalNAc-Ts (GaINAc (N-acetylgalactosamine)-transferases).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85135.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanne Chia et al.
eLife, 3, e01828-e01828 (2014-03-13)
ER O-glycosylation can be induced through relocalisation GalNAc-Transferases from the Golgi. This process markedly stimulates cell migration and is constitutively activated in more than 60% of breast carcinomas. How this activation is achieved remains unclear. Here, we screened 948 signalling
Iva V Klevernic et al.
FEBS letters, 583(4), 680-684 (2009-01-27)
We have investigated the agonists that activate transfected extracellular signal-regulated kinase 8 (ERK8) in cells, and have found that the most potent activators are hydrogen peroxide, DNA alkylating and cross-linking agents and the poly (ADP-ribose) polymerase inhibitor KU-0058948. The feature
Angela L Groehler et al.
The Journal of cell biology, 190(4), 575-586 (2010-08-25)
Proliferating cell nuclear antigen (PCNA) acts as a scaffold, coordinator, and stimulator of numerous processes required for faithful transmission of genetic information. Maintaining PCNA levels above a critical threshold is essential, but little is known about PCNA protein turnover. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service