Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA001174

Sigma-Aldrich

Anti-PIGA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GPI3, Anti-phosphatidylinositol glycan anchor biosynthesis, class A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

FADVSSVLTNKLLTVSLCDTNHIICVSYTSKENTVLRAALNPEIVSVIPNAVDPTDFTPDPFRRHDSITIVVVSRLVYRKGIDLLSGIIPELCQKYPDLNFIIGGEGPKRII

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIGA(5277)

General description

PIGA (phosphatidylinositol glycan anchor biosynthesis, class A) gene consists of six exons and is 17kb in length. It is mapped to chromosome Xp22.1.

Immunogen

phosphatidylinositol glycan anchor biosynthesis, class A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PIGA (phosphatidylinositol glycan anchor biosynthesis, class A) gene encodes a protein that is involved in N-acetylglucosaminyl phosphatidylinositol (GlcNAc-PI) synthesis. GlcNAc-PI is the first intermediate in the biosynthetic pathway of GPI anchor. GPI anchor is a glycolipid that anchors proteins to the cell surface. It is found on RBCs. Mutations in this gene cause paroxysmal nocturnal hemoglobinuria, an acquired hematologic disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74035

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Miyata et al.
Science (New York, N.Y.), 259(5099), 1318-1320 (1993-02-26)
The glycosylphosphatidylinositol (GPI) anchor is a membrane attachment structure of many proteins and occurs in a wide variety of eukaryotes from yeasts to mammals. The structure of the core of the GPI anchor is conserved in protozoa and mammals and
M Bessler et al.
Human molecular genetics, 3(5), 751-757 (1994-05-01)
The PIG-A gene, whose product is involved in one of the early steps in the synthesis of glycan phosphatidylinositol (GPI) anchors, has been recently found to be defective in all cases of paroxysmal nocturnal haemoglobinuria (PNH). By isolating genomic clones
M Bessler et al.
The EMBO journal, 13(1), 110-117 (1994-01-01)
Paroxysmal nocturnal haemoglobinuria (PNH), an acquired clonal blood disorder, is caused by the absence of glycosyl phosphatidylinositol (GPI)-anchored surface proteins due to a defect in a specific step of GPI-anchor synthesis. The cDNA of the X-linked gene, PIG-A, which encodes
J Takeda et al.
Cell, 73(4), 703-711 (1993-05-21)
Paroxysmal nocturnal hemoglobinuria is an acquired hematopoietic disease characterized by abnormal blood cell populations in which the biosynthesis of the glycosylphosphatidylinositol (GPI) anchor is deficient. Deficiency of surface expressions of GPI-anchored complement inhibitors leads to complement-mediated hemolysis. Here we report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service