Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA000841

Sigma-Aldrich

ANTI-TXLNG antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CXorf15, Anti-FIAT, Anti-FLJ11209, Anti-LSR5, Anti-MGC126621, Anti-MGC126625, Anti-TXLNGX

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VSPAYCTQESREEIPGGEARTDPPDGQQDSECNRNKEKTLGKEVLLLMQALNTLSTPEEKLAALCKKYADLLEESRSVQKQMKILQKKQAQIVKEKVHLQSEHSKAILARSKLESLCRELQ

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXorf15(55787)

Looking for similar products? Visit Product Comparison Guide

General description

γ-taxilin (TXLNG) is ubiquitously expressed and belongs to the taxilin family. It interactes preferentially with syntaxin-4. It is found that taxilin family members mutually interact with the syntaxin family members.

Immunogen

γ-Taxilin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

γ-taxilin (TXLNG) is a protein in the cytoplasm that plays an important role in regulating cell cycle. It also plays a role in vesicle trafficking. The gene regulates bone mass accrual and acts as a novel transcriptional regulator of osteoblastic function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74131

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kejun Du et al.
Toxicology, 243(3), 303-310 (2007-12-11)
Lipopolysaccharide (LPS) is the toxic determinant for Gram-negative bacterium infection. The individual response to LPS was related to its gene background. It is necessary to identify new molecules and signaling transduction pathways about LPS. The present study was undertaken to
Vionnie W C Yu et al.
The Journal of cell biology, 169(4), 591-601 (2005-05-25)
We report the characterization of factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription (FIAT), a leucine zipper nuclear protein. FIAT interacted with ATF4 to inhibit binding of ATF4 to DNA and block ATF4-mediated transcription of the osteocalcin gene in vitro.
Satoru Nogami et al.
Biochemical and biophysical research communications, 319(3), 936-943 (2004-06-09)
The syntaxin family is implicated in intracellular vesicle traffic. We have recently identified taxilin, a novel syntaxin-binding protein, which has a long coiled-coil region in its C-terminal half. A database search has revealed the presence of two other molecules having

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service