Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA000728

Sigma-Aldrich

Anti-UPB1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-β-Alanine synthase antibody produced in rabbit, Anti-β-Ureidopropionase antibody produced in rabbit, Anti-BUP-1 antibody produced in rabbit, Anti-N-Carbamoyl-β-alanine amidohydrolase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GALSESLWPIEARNAAIANHCFTCAINRVGTEHFPNEFTSGDGKKAHQDFGYFYGSSYVAAPDSSRTPGLSRSRDGLLVAKLDLNLCQQVNDVWNFKMTGRYEMYARELAEAVKSNYS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UPB1(51733)

Immunogen

β-Ureidopropionase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

β-ureidopropionase is an enzyme encoded by the UPB1 gene in humans. It is the third enzyme of the pyrimidine degradation pathway and catalyzes the conversion of N-carbamyl-β-alanine and N-carbamyl-β-aminoisobutyric acid to β-alanine and β-aminoisobutyric acid, ammonia and CO2. The enzyme catalyzes the final step in the 5-FU catabolic pathway. The gene variants may contribute to the development of 5-FU-related toxicity in some fluoropyrimidine (FP)-treated patients. The gene deficiency is an inborn error of the pyrimidine degradation pathway and affects the cleavage of N-carbamyl-β-alanine and N-carbamyl-β-aminoisobutyric acid.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73418

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cankut Cubuk et al.
Cancer research, 78(21), 6059-6072 (2018-08-24)
Metabolic reprogramming plays an important role in cancer development and progression and is a well-established hallmark of cancer. Despite its inherent complexity, cellular metabolism can be decomposed into functional modules that represent fundamental metabolic processes. Here, we performed a pan-cancer
Cankut Çubuk et al.
NPJ systems biology and applications, 5, 7-7 (2019-03-12)
In spite of the increasing availability of genomic and transcriptomic data, there is still a gap between the detection of perturbations in gene expression and the understanding of their contribution to the molecular mechanisms that ultimately account for the phenotype
A B P van Kuilenburg et al.
Nucleosides, nucleotides & nucleic acids, 25(9-11), 1093-1098 (2006-10-27)
beta-Ureidopropionase is the third enzyme of the pyrimidine degradation pathway and it catalyses the irreversible hydrolysis of N-carbamyl-ss-aminoisobutyric acid or N-carbamyl-ss-alanine to beta-aminoisobutyric acid or ss-alanine, ammonia, and CO2. Analysis of the beta-ureidopropionase gene (UPB1) of the first 4 patients
Julie Fidlerova et al.
Pharmacological reports : PR, 64(5), 1234-1242 (2012-12-15)
An impairment of the 5-fluorouracil (5-FU) catabolic pathway, represented by alterations in the dihydropyrimidine dehydrogenase (DPYD) gene, is considered a crucial factor contributing to the development of 5-FU-related toxicity. The β-ureidopropionase (BUP1) enzyme catalyzes the final step in the 5-FU
André B P van Kuilenburg et al.
Biochimica et biophysica acta, 1822(7), 1096-1108 (2012-04-25)
ß-ureidopropionase is the third enzyme of the pyrimidine degradation pathway and catalyzes the conversion of N-carbamyl-ß-alanine and N-carbamyl-ß-aminoisobutyric acid to ß-alanine and ß-aminoisobutyric acid, ammonia and CO(2). To date, only five genetically confirmed patients with a complete ß-ureidopropionase deficiency have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service