Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA000638

Sigma-Aldrich

Anti-ARFGAP3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ADP-ribosylation factor GTPase-activating protein 3 antibody produced in rabbit, Anti-ARF GAP 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SKQDILTIFKRLRSVPTNKVCFDCGAKNPSWASITYGVFLCIDCSGSHRSLGVHLSFIRSTELDSNWSWFQLRCMQVGGNASASSFFHQHGCSTNDTNAKYNSRAAQLYREKIKSLASQATRKHGTDLWLDSCVVPPLSPPP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARFGAP3(26286)

General description

ADP-ribosylation factor GTPase-activating protein 3 is an enzyme encoded by the ARFGAP3 gene in humans and belongs to ARF GAP family. It is a predominantly cytosolic protein and is concentrated in the perinuclear region.

Immunogen

ADP-ribosylation factor GTPase-activating protein 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-ARFGAP3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ADP-ribosylation factor GTPase-activating protein 3 acts as a novel GTPase-activating protein (GAPs) for ADP ribosylation factors 1 (ARF1) and may be associated with intracellular trafficing of proteins and vesicular transport. The protein is associated with the Golgi apparatus and regulates the vesicular trafficking pathway. It is also considered as a novel androgen-regulated gene that can promote prostate cancer cell proliferation and migration in collaboration with paxillin. It is found to be strongly expressed in endocrine glands and testis. Its expression in testis is about sixfold higher than that in ovary and may have a possible role in the physiological function of sperm. Its expression is also observed in four human fetal tissues and seven cancer cell lines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73439

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Zhang et al.
Genomics, 63(3), 400-408 (2000-03-08)
We have identified and characterized a novel human ADP-ribosylation factor GTPase-activating protein (ARFGAP1) gene that is related to other members of the ARF GAP family. The full-length cDNA for human ARFGAP1 was cloned following the identification of an EST obtained
Daisuke Obinata et al.
International journal of cancer, 130(10), 2240-2248 (2011-06-08)
ADP ribosylation factor GTPase-activating protein 3 (ARFGAP3) is a GTPase-activating protein that associates with the Golgi apparatus and regulates the vesicular trafficking pathway. In the present study, we examined the contribution of ARFGAP3 to prostate cancer cell biology. We showed
X Liu et al.
FEBS letters, 490(1-2), 79-83 (2001-02-15)
ADP ribosylation factors (ARFs) are critical in the vesicular trafficking pathway. ARF activity is controlled by GTPase-activating proteins (GAPs). We have identified recently a novel tentative ARF GAP derived from human fetal liver, ARFGAP3 (originally named as ARFGAP1). In the
Mayur Dembla et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(15), 5245-5260 (2014-04-11)
Ribbon synapses are tonically active synapses in the retina and inner ear with intense vesicle traffic. How this traffic is organized and regulated is still unknown. Synaptic ribbons, large presynaptic structures associated with numerous synaptic vesicles, appear to be essential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service