Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HMI0021

Sigma-Aldrich

MISSION® microRNA Mimic

hsa-let-7i

Synonym(s):

hsa-let-7i-5p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

product line

MISSION®

form

solid

mature sequence

UGAGGUAGUAGUUUGUGCUGUU

Sanger mature/minor accession no.

Sanger microRNA accession no.

storage temp.

−20°C

Gene Information

General description

The ready-to-use MISSION miRNA mimics are small, double-stranded RNA molecules designed to mimic endogenous mature miRNA molecules when introduced into cells. miRNA are known to regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation. MISSION miRNA Mimics, a member of MISSION RNAi product family, provides miRNA researchers with a range of options from individual MISSION mimics to a full library of human miRNA mimics based on latest version of miRBase (currently hosted by the University of Manchester, previously hosted by the Sanger Institute).

  • Optimized and ready for transfection.
  • Novel MISSION miRNA mimic design has been functionally tested for knockdown efficiency against natural miRNA targets.
  • Unique MISSION miRNA mimic design significantly reduces possible sense strand off target effects.
  • Available as a whole human library and individual miRNA targets.

Biochem/physiol Actions

The miRNA hsa-let-7i-5p is associated with colorectal cancer metastasis. Its targets include TRIM41 (tripartite motif-containing protein 41), SOX13 (SRY (sex determining region Y)-box 13), SLC25A4 (solute carrier family 25 member 4), SEMA4F (semaphorin-4F), RPUSD2 (RNA pseudouridylate synthase domain-containing protein 2), PLEKHG6 (pleckstrin homology domain-containing family G member 6), CCND2 (G1/S-specific cyclin-D2) and BTBD3 (BTB/POZ domain-containing protein 3).[1] It also targets TLR4 (toll like receptor) and can inhibit TLR4-mediated damage.[2]

Other Notes

miRBase V18 Mature ID update: hsa-let-7i-5p

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive gene and microRNA expression profiling reveals the crucial role of hsa-let-7i and its target genes in colorectal cancer metastasis.
Zhang P
Molecular Biology Reports, 39, 1471-1471 (2012)
Hsa-mir-let 7i Treatment Suppresses Tlr4 Mediated Mitochondrial Oxidative Stress In Giant Cell Arteritis.
Rao NA and Saraswathy S
Investigative Ophthalmology & Visual Science, 53, 6243-6243 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service