Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H9915

Sigma-Aldrich

Anti-HOXB6 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HOX-2B, Anti-HOX2.2, Anti-HU-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using using lysates of HEK-293T cells over expressing human HOXB6

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXB6(3216)
mouse ... Hoxb6(15414)

General description

Homeobox 6 (HOXB6) protein is expressed in human hematopoietic progenitor cells (HPCs). The HOXB6 gene is mapped on the human chromosome at 17q21.32.

Specificity

Anti-HOXB6 (C-terminal) recognizes human HOXB6 (also known as HOX2B, HU-2 and Hox-2.

Immunogen

synthetic peptide corresponding to amino acids 108-122 of human HOXB6 conjugated to KLH. The corresponding sequence is identical in mouse.

Application

Anti-HOXB6 (C-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Homeobox 6 (HOXB6) is frequently overexpressed in human acute myeloid leukemia (AML). HOXB6 gene overexpression extended hematopoietic stem cells (HSCs) and myeloid precursors while decreasing erythropoiesis and lymphopoiesis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression pattern of HOXB6 homeobox gene in myelomonocytic differentiation and acute myeloid leukemia
Giampaolo A, et al.
Leukemia, 16(7), 1293-1301 (2002)
Genomic alterations in lobular neoplasia: a microarray comparative genomic hybridization signature for early neoplastic proliferationin the breast
Mastracci TL, et al.
Genes Chromosomes Cancer, 45(11), 1007-1017 (2006)
Neal A Fischbach et al.
Blood, 105(4), 1456-1466 (2004-11-04)
The HOX family of homeobox genes plays an important role in normal and malignant hematopoiesis. Dysregulated HOX gene expression profoundly effects the proliferation and differentiation of hematopoietic stem cells (HSCs) and committed progenitors, and aberrant activation of HOX genes is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service