Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H9913

Millipore

HIS-Select® iLAP 5 mL Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

description

Disposable gravity column

capacity

≥1 mg binding capacity (histidine-tagged protein from a 5 mL bacterial culture)

storage temp.

2-8°C

Application

The HIS-Select iLAP (Integrated Lysis and Affinity Purification) Columns combine cell lysis and protein purification steps into one. These single-use, disposable columns are designed for the one-step purification of histidine-tagged proteins directly from a 5 mL bacterial culture. The single-step method uses Sigma′s patented iLAP technology, which allows quick and simple histidine-tagged protein purification from recombinant clones. Each column contains nickel chelate resin for the purification, as well as five lysis reagent tablets, which include all the necessary detergents and enzymes needed for efficient cell lysis and protein extraction while eliminating the need to harvest the cells from culture. Each column is capable of purifying at least 1 mg of histidine-tagged protein directly from 5 mL of bacterial culture in less than one hour. The procedure provided can be used to extract soluble proteins directly from growing cells and results in a nearly pure fusion protein preparation following elution from the column. This makes these columns ideal for confirming expression of a histidine-tagged target protein, for performing protein-protein interaction assays, or for rapidly screening colonies or multiple constructs (cultures). The resulting purified protein is compatible with protein assays, SDS-PAGE, Western blotting, and MALDI.

Legal Information

HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany
iLAP is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • C5491Cellytic Express, 1 mL Tablets, 1 tablet sufficient for 1 mL lysis (cell suspension)SDS

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sivakamasundari Pichu et al.
BMC biochemistry, 12, 32-32 (2011-06-29)
Ixodes scapularis, commonly known as the blacklegged or deer tick, is the main vector of Lyme disease in the United States. Recent progress in transcriptome research has uncovered hundreds of different proteins expressed in the salivary glands of hard ticks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service