H9776
Heat Shock Protein 70 from bovine brain
>95% (SDS-PAGE), lyophilized powder
Synonym(s):
HSP 70
About This Item
Recommended Products
biological source
bovine brain
assay
>95% (SDS-PAGE)
form
lyophilized powder
composition
Protein, ~10% Lowry
technique(s)
western blot: suitable
storage temp.
−20°C
Gene Information
bovine ... HSPA1A(282254) , HSPA8(281831)
Application
Biochem/physiol Actions
Physical form
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service