Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H9534

Sigma-Aldrich

Hepatitis C virus NS4 antigen

~1 mg/mL, ≥95% (SDS-PAGE), recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

assay

≥95% (SDS-PAGE)

mol wt

19 kDa (plus 114 kDa β-Gal tag)

concentration

~1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

hepatitis C virus ... HCVgp1(951475)

Biochem/physiol Actions

Positive control for HCV antibodies.

Other Notes

HCV NS4a+b [1658-1863]-Galactosidase-tagged

Physical form

Solution in 8 M urea, 20 mM tris-HCl, pH 8.0, and 10 mM 2-mercaptoethanol.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Bartenschlager et al.
Journal of virology, 69(12), 7519-7528 (1995-12-01)
Processing of the hepatitis C virus polyprotein is mediated by host cell signalases and at least two virally encoded proteinases. Of these, the serine-type proteinase encompassing the amino-terminal one-third of NS3 is responsible for cleavage at the four sites carboxy
Miriam T Brady et al.
European journal of immunology, 33(12), 3448-3457 (2003-11-25)
The majority of hepatitis C virus (HCV) infections become chronic, despite the presence of HCV-specific cellular and humoral immune responses. We have previously suggested that IL-10-secreting antigen-specific regulatory T cells may contribute to viral persistence, and demonstrate here that peripheral
Inés Romero-Brey et al.
mBio, 6(4), e00759-e00759 (2015-07-15)
Induction of membrane rearrangements in the cytoplasm of infected cells is a hallmark of positive-strand RNA viruses. These altered membranes serve as scaffolds for the assembly of viral replication factories (RFs). We have recently shown that hepatitis C virus (HCV)
Margaret A Scull et al.
PLoS pathogens, 11(11), e1005297-e1005297 (2015-11-21)
The hepatitis C virus (HCV) p7 protein is required for infectious virus production via its role in assembly and ion channel activity. Although NMR structures of p7 have been reported, the location of secondary structural elements and orientation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service