Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H8756

Sigma-Aldrich

Hemoglobin–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817

biological source

protein from bovine

form

saline suspension

extent of labeling

>6 mg per mL

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Villarreal et al.
Applied and environmental microbiology, 74(18), 5854-5856 (2008-08-05)
To produce recombinant hemoglobin in Escherichia coli, sufficient intracellular heme must be present, or the protein folds improperly and is degraded. In this study, coexpression of human hemoglobin genes and Plesiomonas shigelloides heme transport genes enhanced recombinant hemoglobin production in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service