Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H8158

Sigma-Aldrich

Heat Shock Protein 27 human

recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

HSP 27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

Quality Level

recombinant

expressed in E. coli

assay

~90% (SDS-GE)

form

buffered aqueous solution

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HSPB1(3315)

Biochem/physiol Actions

Heat shock proteins (HSPs) are molecular chaperones. They prevent aggregation of unfolded proteins. HSP27 is involved in anti-apoptosis and actin, and microtubule stabilization. It exists in unphosphorylated inactive form and phosphorylated active form.[1] In monocytes, phosphorylated HSP27 is associated with bone mineral density and can contribute to the pathogenesis of osteoporosis.[2] It also participates in iron metabolism and negatively regulates ferroptosis-associated cancer cell death.[3]

Physical form

Solution in 20 mM Tris HCl, pH 7.5, 10 mM NaCl, 1 mM EDTA, 1 mM DTT

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haruhiko Tokuda et al.
PloS one, 10(6), e0128977-e0128977 (2015-06-06)
We investigated the relationship between HSP27 phosphorylation and collagen-stimulated activation of platelets in patients with diabetes mellitus (DM). Platelet-rich plasma was prepared from blood of type 2 DM patients. The platelet aggregation was analyzed in size of aggregates by an
Bhavna Daswani et al.
Disease markers, 2015, 196589-196589 (2015-06-13)
Peripheral monocytes, precursors of osteoclasts, have emerged as important candidates for identifying proteins relevant to osteoporosis, a condition characterized by low Bone Mineral Density (BMD) and increased susceptibility for fractures. We employed 4-plex iTRAQ (isobaric tags for relative and absolute
K Engel et al.
Biomedica biochimica acta, 50(9), 1065-1071 (1991-01-01)
A hybrid protein containing the N-terminal part of the murine stress protein hsp25 (amino acids 1 to 110) and the C-terminal part of the human stress protein hsp27 (amino acids 111 to 208) was expressed in E. coli using a
X Sun et al.
Oncogene, 34(45), 5617-5625 (2015-03-03)
Ferroptosis is an iron-dependent form of non-apoptotic cell death, but its molecular mechanism remains largely unknown. Here, we demonstrate that heat shock protein beta-1 (HSPB1) is a negative regulator of ferroptotic cancer cell death. Erastin, a specific ferroptosis-inducing compound, stimulates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service