Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

H7287

Sigma-Aldrich

Anti-Heat Shock Protein 110 (HD-19) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Hsp110

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 110 kDa

species reactivity

mouse, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 2-4 μg/mL using rat cerebellum
microarray: suitable
western blot: 0.5-1 μg/mL using cytosolic fraction of rat brain and whole cell extract of mouse fibroblast NIH3T3 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Hsph1(15505)

General description

Heat shock protein 110 (Hsp110) belongs to a family of large stress proteins referred to as the SSE family. This protein is a normal constituent of mammalian cells, ubiquitously expressed but at higher levels in the brain.

Specificity

Anti-Heat Shock Protein 110 (Hsp110) (HD-19) recognizes Hsp110 (110 kDa, may appear as a doublet band in some cell/tissue extracts).

Immunogen

synthetic peptide located at the C-terminus of mouse Hsp110 (amino acids 840-858), conjugated to KLH.

Application

Anti-Heat Shock Protein 110 (HD-19) antibody produced in rabbit may be used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Heat shock protein 110 (Hsp110) is associated with the nucleoli of non-stressed and heat-stressed murine cells. Overexpression of Hsp110 gene has been shown to confer cellular heat resistance to prevent protein aggregation and to keep denatured protein in a folding-competent state. HSP110 plays a role in the pathophysiology of brain damage such as ischemia/reperfusion injuries and pathological conditions such as Alzheimer′s disease.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For prolonged storage, freeze in working aliquots at −20 °C. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binnur Eroglu et al.
Molecular and cellular biology, 30(19), 4626-4643 (2010-08-04)
Accumulation of tau into neurofibrillary tangles is a pathological consequence of Alzheimer's disease and other tauopathies. Failures of the quality control mechanisms by the heat shock proteins (Hsps) positively correlate with the appearance of such neurodegenerative diseases. However, in vivo
B L Hylander et al.
Brain research, 869(1-2), 49-55 (2000-06-24)
Hsp110 is one of the few, major heat shock proteins of mammalian cells and was one of the earliest heat shock proteins described. However, it has only recently been cloned and studied at the molecular level. It has been noted
H J Oh et al.
The Journal of biological chemistry, 272(50), 31636-31640 (1998-02-12)
The 110-kDa heat shock protein (hsp110) has long been recognized as one of the primary heat shock proteins in mammalian cells. It belongs to a recently described protein family that is a significantly diverged subgroup of the hsp70 family and
D P Easton et al.
Cell stress & chaperones, 5(4), 276-290 (2000-10-26)
Both the Grp170 and Hsp110 families represent relatively conserved and distinct sets of stress proteins, within a more diverse category that also includes the Hsp70s. All of these families are found in a wide variety of organisms from yeasts to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service