Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H7035

Sigma-Aldrich

Hepatitis C virus core antigen−Biotin

~1 mg/mL, >95% (SDS-PAGE), recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

recombinant

expressed in E. coli

assay

>95% (SDS-PAGE)

mol wt

22 kDa (plus 114 β-Gal tag)

concentration

~1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

hepatitis C virus ... HCVgp1(951475)

Biochem/physiol Actions

Positive control for HCV antibodies.

Other Notes

HCV core antigen [2-192]-Galactosidase-tagged.

Physical form

Solution in 8 M Urea, 20 mM Tris-HCl, pH 8.0.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service