Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6910

Sigma-Aldrich

13S-Hydroxy-9Z,11E-octadecadiene-(2-biotinyl)hydrazide

≥98%, ethanol solution

Synonym(s):

13(S)-HODE−biotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28N4O4S
Molecular Weight:
516.61
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Quality Level

assay

≥98%

form

ethanol solution

impurities

biotin, may contain traces

εmax

30,000 at 234 nm

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\C=C/CCCCCCCC(=O)NNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12

InChI

1S/C28H48N4O4S/c1-2-3-11-16-22(33)17-12-9-7-5-4-6-8-10-13-19-25(34)31-32-26(35)20-15-14-18-24-27-23(21-37-24)29-28(36)30-27/h7,9,12,17,22-24,27,33H,2-6,8,10-11,13-16,18-21H2,1H3,(H,31,34)(H,32,35)(H2,29,30,36)/b9-7-,17-12+/t22-,23-,24-,27-/m0/s1

InChI key

VGSREJALHGALEV-YJERLKHQSA-N

Application

May be used as a probe to detect binding proteins and/or receptors for 13(S)-HODE.

Biochem/physiol Actions

Modulates the platelet-activating factor, leukotriene B4, and formyl-Met-Leu-Phe-induced calcium influx in human polymorphonuclear leukocytes.

Other Notes

13(S)-HODE is the lipoxygenase metabolite of linoleic acid.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service