Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6270

Sterilizer replacement heater element

AC/DC input 240 V AC

Synonym(s):

replacement element, replacement heating elements

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161502
NACRES:
NB.22

description

For use with Sterilizer, infrared heat

availability

not available in EU (Not available in EU countries)

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Contains no asbestos

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service