Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6146

Sigma-Aldrich

Methyl 2-hydroxybehenate

≥98% (capillary GC)

Synonym(s):

DL-α-Hydroxybehenic acid methyl ester, Methyl 2-hydroxydocosanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46O3
CAS Number:
Molecular Weight:
370.61
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥98% (capillary GC)

form

solid

functional group

ester

Storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCC(O)C(=O)OC

InChI

1S/C23H46O3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(24)23(25)26-2/h22,24H,3-21H2,1-2H3

Inchi Key

KUHHQOLYDNMSJW-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fereshteh Farokhi et al.
Marine drugs, 8(12), 2988-2998 (2011-02-23)
The starfish Narcissia canariensis harvested from the coasts off Dakar, Senegal, was investigated for glycolipids (GL). This report deals with the isolation, characterization and biological activity of a fraction F13-3 separated from the GL mixture and selected according to its
M V Levitina
Zhurnal evoliutsionnoi biokhimii i fiziologii, 20(4), 347-352 (1984-07-01)
Brain cerebrosides (C) and sulfocerebrosides (S) of Insectivora, which represent the most ancient and primitive order of placentary mammals, were first studied. The content of C and S is higher in hedgehogs (Erinaceus europaeus, E. auritus) and mole (Talpa europaea)
Nina Blaas et al.
Journal of agricultural and food chemistry, 59(11), 6018-6024 (2011-05-04)
The sphingolipid composition of food as well as of physiological samples has received considerable interest due to their positive biological activities. This study quantified the total amount of sphingomyelin (SM) in 20 human breast milk samples from healthy volunteers and
Jin-Ming Gao et al.
Chemistry and physics of lipids, 131(2), 205-213 (2004-09-08)
The ceramide fractions were isolated from the chloroform/methanolic extractable of the fruiting bodies of Tuber indicum and separated into three kinds of molecular species TI-1, TI-2, and TI-3 by normal and reverse phase silica gel-column chromatography. By means of (1)H
M Bohn et al.
Archives of biochemistry and biophysics, 387(1), 35-40 (2001-05-23)
Although the results of lipid analyses from several plant species have been available for many years a complete characterization of the corn root plasma membrane is still lacking. The present study provides a detailed analysis of individual lipids and a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service