H6020
Hybridoma Enhancing Supplement Hybri-Max™ from murine thymoma cell line
liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Synonym(s):
Conditioned Medium from murine thymoma cell line
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352205
Recommended Products
sterility
sterile-filtered
Quality Level
form
liquid
technique(s)
cell culture | hybridoma: suitable
cell culture | mammalian: suitable
impurities
Mycoplasma, tested
endotoxin, tested
shipped in
dry ice
storage temp.
−20°C
Application
Use at a concentration of 10% in complete medium containing FBS (10%) or CPSR-3. Optimal conditions must be determined for specific applications.
Other Notes
Conditioned medium from a proprietary murine thymoma cell line grown in RPMI-1640 with 15% FBS.
Analysis Note
Cell line virus testing:
Murine Antibody Production (MAP) testing for Sendai, MHV, PVM, Reo 3, GDVII, Ectomelia, MVM, Polyoma, LDVM, M. Ad. EDIM MCMV, LCM, Hantaan.
Murine Antibody Production (MAP) testing for Sendai, MHV, PVM, Reo 3, GDVII, Ectomelia, MVM, Polyoma, LDVM, M. Ad. EDIM MCMV, LCM, Hantaan.
Legal Information
Hybri-Max is a trademark of Sigma-Aldrich Co. LLC
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service