Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H5899

Sigma-Aldrich

Hanks′ Balanced Salts

Without phenol red and sodium bicarbonate, liquid, sterile-filtered

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207

sterility

sterile-filtered

form

liquid

Looking for similar products? Visit Product Comparison Guide

General description

Formulation: click here.

Application

For use in Mycoplasma Stain Kit.

Other Notes

The pH is adjusted after addition of sodium bicarbonate. Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when working with stem cells or when growing cells at low densities.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service