Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H5291

Sigma-Aldrich

Fibroblast Growth Factor-8b human

recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

15-60 ng/mL ED50

mol wt

dimer 30-45 kDa (glycosylated)

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

−20°C

Biochem/physiol Actions

FGF-8b belongs to the FGF protein family. FGF-8b is important in embryogenesis, in pathways that lead to left-right determination and coordinate the vertebral segmentation process, and in processes that activate Homeobox genes. There are several isoforms of FGF-8 produced by alternative splicing.

Preparation Note

HumanKine Fibroblast Growth Factor-8b, expressed in human HEK 293 cells is a glycosylated monomer with an apparent molecular mass of 30-45 kDa due to glycosylation. Production in human HEK 293 cells offers authentic glycosylation, which is absent when this cytokine is expressed in E. coli. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The specific activity was determined by the dose-dependent stimulation of the proliferation of the BALB/3T3 cell line.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maija P Valta et al.
Journal of cellular biochemistry, 107(4), 769-784 (2009-05-06)
Fibroblast growth factor 8 (FGF-8) is expressed at an increased level in a high proportion of prostate cancers and it is associated with a poor prognosis of the disease. Our aim was to study the effects of FGF-8b on proliferation

Articles

Fibroblast growth factors in cell culture and various growth factors for your research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service