Sigma-Aldrich
diam.
3 1/8 in. , hole
General description
Keeps squeeze and other bottles from tipping over as bottle becomes top heavy. Units feature 3 1/8 in. or 3 3/4 in. ID holes and mount instantly with 3-M double-faced tape. Can be screw-mounted on wall, hood or secured to benchtop.
Saves on solvents and prevents spills. Useful next to sinks or other solvent wash areas.
Saves on solvents and prevents spills. Useful next to sinks or other solvent wash areas.
Choose from one of the most recent versions:
Certificates of Analysis (COA)