H3894
8(R)-Hydroxy-(5Z,9E,11Z,14Z)-eicosatetraenoic acid
~100 μg/mL in ethanol, ~98%
Synonym(s):
8(R)-HETE
About This Item
assay
~98%
form
liquid
concentration
~100 μg/mL in ethanol
functional group
carboxylic acid
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCC\C=C/C/C=C\C=C\[C@H](O)C\C=C/CCCC(O)=O
InChI
1S/C20H32O3/c1-2-3-4-5-6-7-8-9-10-13-16-19(21)17-14-11-12-15-18-20(22)23/h6-7,9-11,13-14,16,19,21H,2-5,8,12,15,17-18H2,1H3,(H,22,23)/b7-6-,10-9-,14-11-,16-13+/t19-/m0/s1
InChI key
NLUNAYAEIJYXRB-GTYUHVKWSA-N
Packaging
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
57.2 °F - closed cup - Solvent
flash_point_c
14.0 °C - closed cup - Solvent
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service