Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H2271

Sigma-Aldrich

[Ser 442,447]-Heat Shock Protein 65, fragment 437-450

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.26

Product Name

[Ser 442,447]-Heat Shock Protein 65, fragment 437-450, ≥97% (HPLC)

assay

≥97% (HPLC)

form

lyophilized powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HSPD1(3329)

Biochem/physiol Actions

Protects against neurotoxins related to HIV infection and Alzheimer′s disease

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Brenneman et al.
The Journal of clinical investigation, 97(10), 2299-2307 (1996-05-15)
A novel 14-amino acid peptide, with stress-protein-like sequences, exhibiting neuroprotection at unprecedented concentrations, is revealed. This peptide prevented neuronal cell death associated with the envelope protein (GP 120) from HIV, with excitotoxicity (N-methyl d-aspartate), with the beta amyloid peptide (putative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service