Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H2161

Sigma-Aldrich

8-Hydroxyadenosine-3′,5′-monophosphorothioate, Rp-isomer

>99% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O6PSNa
Molecular Weight:
383.25
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

assay

>99% (HPLC)

form

solid

mol wt

390.26

packaging

vial of 1 μmol (approx. 0.4mg)

storage temp.

−20°C

SMILES string

Nc1ncnc2n([C@@H]3O[C@@H]4CO[P@@](S)(=O)O[C@H]4[C@H]3O)c(O)nc12

InChI

1S/C10H12N5O6PS/c11-7-4-8(13-2-12-7)15(10(17)14-4)9-5(16)6-3(20-9)1-19-22(18,23)21-6/h2-3,5-6,9,16H,1H2,(H,14,17)(H,18,23)(H2,11,12,13)/t3-,5-,6-,9-,22-/m1/s1

InChI key

ZJLRJSRRXWDPOZ-KVBUDSETSA-N

Biochem/physiol Actions

Phosphodiesterase-resistant, polar, membrane-impermeable inhibitor of cAMP-dependent protein kinase (PKA).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Puzenko et al.
The Journal of chemical physics, 137(19), 194502-194502 (2012-11-28)
In this, the third part of our series on the dielectric spectrum symmetrical broadening of water, we consider the nucleotide aqueous solutions. Where in Parts I [E. Levy et al., J. Chem. Phys. 136, 114502 (2012)] and II [E. Levy
B T Gjertsen et al.
The Journal of biological chemistry, 270(35), 20599-20607 (1995-09-01)
Novel (Rp)-cAMPS analogs differed widely in ability to antagonize cAMP activation of pure cAMP-dependent protein kinase I and II and to antagonize actions of cAMP on gene expression, shape change, apoptosis, DNA replication, and protein phosphorylation in intact cells. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service