Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H2132

Sigma-Aldrich

DL-Hexanoylcarnitine chloride

Synonym(s):

DL-Caproylcarnitine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H26NO4Cl
CAS Number:
Molecular Weight:
295.80
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

lipid type

saturated FAs

storage temp.

−20°C

SMILES string

Cl.CCCCCC(=O)OC(CC(O)=O)C[N](C)(C)C

InChI

1S/C13H26NO4.ClH/c1-5-6-7-8-13(17)18-11(9-12(15)16)10-14(2,3)4;/h11H,5-10H2,1-4H3,(H,15,16);1H

InChI key

AWUOLOICNWRXPY-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Schmidt-Sommerfeld et al.
Pediatric research, 31(6), 545-551 (1992-06-01)
To determine the sensitivity and specificity of detecting urinary medium-chain acylcarnitines for the diagnosis of MCAD deficiency, 114 urine specimens from 75 children with metabolic diseases and controls were analyzed in a blinded fashion using a radioisotopic exchange/HPLC method. All
A K Bhuiyan et al.
Clinical and investigative medicine. Medecine clinique et experimentale, 18(2), 144-151 (1995-04-01)
A sensitive method of continuous on-line radio-high performance liquid chromatography (HPLC) was used to detect the specific radio-labelled acyl-carnitine esters derived from the oxidation of [U-14C]3-methyl-2-oxopentanoate by rat liver and muscle mitochondrial fractions. The recoveries of carnitine, acetyl-carnitine, propionyl-carnitine, 2-methylbutyryl-carnitine
H Caksen et al.
Genetic counseling (Geneva, Switzerland), 15(1), 9-17 (2004-04-16)
Lissencephaly or agyria refers to a rare disorder that is characterized by the absence of cerebral convolutions and a poorly formed sylvian fissure, giving the appearance of a 3-4 months old fetal brain. At present more than 25 dysmorphology syndromes
Anelise M Tonin et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 30(5), 383-390 (2012-04-05)
Medium-chain fatty acids and acylcarnitines accumulate in medium-chain acyl-CoA dehydrogenase deficiency (MCADD), the most frequent fatty acid oxidation defect clinically characterized by episodic crises with vomiting, seizures and coma. Considering that the pathophysiology of the neurological symptoms observed in MCADD
P E Minkler et al.
Analytical biochemistry, 212(2), 510-518 (1993-08-01)
This paper describes a method for the quantitative determination of free carnitine, acetylcarnitine, propionylcarnitine, hexanoylcarnitine, octanoylcarnitine, and total carnitine in plasma. Carnitine and acylcarnitines were extracted from 100 microliters of plasma with acetonitrile/methanol and isolated using 0.5-ml columns of silica

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service