Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H17003

Sigma-Aldrich

G4 Dendritic Cell Generation Xeno-Free HumanKine® Cytokine Cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

≤0.6 ng/mL IL-4 EC50
≤2.0 ng/mL GM-CSF EC50

mol wt

14-19 kDa (IL-4, monomer, glycosylated)
15-36 kDa (GM-CSF, monomer, glycosylated)

impurities

<1.0 EU/μg endotoxin

storage temp.

−70°C

General description

Produces high quality dendritic cells, Xeno and Animal-derived component free.

Reconstitution

Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in 1X PBS containing 0.1% endotoxin-free recombinant human serum albumin.

Storage and Stability

Upon reconstitution, the cytokine can be stored at 4°C for short term or at -20°C to -80°C in aliquots for long term. Avoid repeated freeze-thaw cycles.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service