LIF promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation.
Reconstitution
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1X PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Storage and Stability
Upon reconstitution, the cytokine can be stored at 4°C for short term only or at -20°C to -80°C in aliquots for long term. Avoid repeated freeze-thaw cycles.
Analysis Note
The activity was determined by the dose- dependent stimulation of the proliferation of human TF-1 cells (human erythroleukemic indicator cell line).
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.