Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H1290

Sigma-Aldrich

HIV Protease Mutant

recombinant, expressed in E. coli

Synonym(s):

(L33F, M46L, I54V, V82A, L90M, L10I, M36I, A71V, L63P)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

solution

mol wt

~10.8 kDa

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Stabilized without non-physiologic mutations

Packaging

100 unit vial containing ≥ 100ug protein

Unit Definition

One unit of HIV protease will cleave 1picomole per minute of the substrate Abz-Ala-Arg-Val-Nle-Tyr(NO2)-Glu-Ala-Nle-NH2 at 37°C.

Physical form

Supplied as a solution in 0.5M Sodium Acetate, pH 4.7with 50mM Tris-HCL, 1M NaCL, 1mM EDTA, 5mM DTT and 15% glycerol.

Preparation Note

Bioengineered to resist autoproteolysis, resulting in an enzyme with improved stability and substantially higher activity levels over extended time periods to conduct kinetic assays. The enzyme includes the most commonly occurring background polymorphism (L63P) that aids in the drug resistance profile.

substrate

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service