Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H1284

Hybri-well Press-seal hybridization chambers

chamber diam. × D 20 mm × 0.15 mm

Synonym(s):

Hydrophobic cover slip, polycarbonate chamber slide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

chamber diam. × D

20 mm × 0.15 mm

volume

30 μL

Looking for similar products? Visit Product Comparison Guide

General description

Hydrophobic cover slip with non-cytotoxic spacer adheres to a microscope slide, forming a chamber with access ports for addition and removal of reagents. RNase-free, heat-resistant, and disposable: ideal for hybridization. Easily removed from microscope slide after hybridization steps.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service