Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H1041

Sigma-Aldrich

Anti-HOXC10 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Homeobox C10, Anti-Homeoprotein C10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysate of HEK-293T cells over-expressing human HOXC10
western blot: 1-2 μg/mL using lysates of HEK-293T cells over-expressing human HOXC10

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXC10(3226)

General description

Homeobox C10 (HOXC10) protein expression is cell-type-dependent. The HOXC10 gene is a member of the HOX gene family. This gene encodes for a protein that consists of conserved DNA binding homeodomain. The HOXC10 gene is mapped on the human chromosome at 12q13.13.

Specificity

Anti-HOXC10 (C-terminal) recognizes human HOXC10.

Application

Anti-HOXC10 (C-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Homeobox C10 (HOXC10) is associated with cell proliferation. HOXC10 is targeted for degradation, early in mitosis by the ubiquitin-proteasome pathway. HOXC10 gene is expressed at higher levels in thyroid cancer, cervical cancer, and lung adenocarcinoma. Upregulation of HOXC10 gene expression is associated with proliferation, invasion, and migration of gliomas. Overexpression of the HOXC10 gene is observed in breast carcinoma.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingzong Li et al.
Stem cells (Dayton, Ohio), 37(2), 247-256 (2018-10-26)
The characteristics of mesenchymal stromal cells (MSCs) which derived from multiple myeloma (MM) patients are typically impaired in osteogenic differentiation. However, the underlying molecular mechanisms need to be further investigated. lncRNAs are emerging as critical regulation molecules in oncogenic pathways.
Helen Sadik et al.
Cancer research, 76(15), 4443-4456 (2016-06-16)
Development of drug resistance is a major factor limiting the continued success of cancer chemotherapy. To overcome drug resistance, understanding the underlying mechanism(s) is essential. We found that HOXC10 is overexpressed in primary carcinomas of the breast, and even more
E de Stanchina et al.
Journal of molecular biology, 299(3), 667-680 (2000-06-03)
We have previously shown that a cell cycle-dependent nucleoprotein complex assembles in vivo on a 74 bp sequence within the human DNA replication origin associated to the Lamin B2 gene. Here, we report the identification, using a one-hybrid screen in
Zhanyao Tan et al.
Theranostics, 8(18), 5143-5158 (2018-11-16)
High levels of angiogenesis are associated with poor prognosis in patients with gliomas. However, the molecular mechanisms underlying tumor angiogenesis remain unclear. Methods: The effect of homeobox C10 (HOXC10) on tube formation, migration, and proliferation of human umbilical vein endothelial
Davide Gabellini et al.
The EMBO journal, 22(14), 3715-3724 (2003-07-11)
Hox proteins are transcription factors involved in controlling axial patterning, leukaemias and hereditary malformations. Here, we show that HOXC10 oscillates in abundance during the cell cycle, being targeted for degradation early in mitosis by the ubiquitin-dependent proteasome pathway. Among abdominal-B

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service