Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0791

Sigma-Aldrich

Anti-HOXA10 (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HOX-1.8, Anti-HOX1H, Anti-Homeobox 1H, Anti-Homeobox A10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:1,000 using lysate of HEK-293T cells over expressing human HOXA10

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXA10(3206)

General description

Homeobox A10 (HOXA10) belongs to the HOX transcription factor family. HOX transcription factors are classified as four clusters, such as cluster A, B, C and D. HOXA10 is localized in A cluster on chromosome 7.

Application

Anti-HOXA10 (C-terminal) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Homeobox A10 (HOXA10) is highly essential for the growth of erythroid and megakaryocytic lineages. The expression of HOXA10 was found to be altered in endometriosis. It also participates in gene expression, morphogenesis and differentiation. HOXA10 plays a key role in gastric cancer, oral squamous cell carcinoma, prostate carcinoma, ovarian clear cell adenocarcinoma, endometrial adenocarcinoma.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LncHOXA10 drives liver TICs self-renewal and tumorigenesis via HOXA10 transcription activation
Shao M, et al.
Molecular Cancer, 17 (2018)
HOXA10 is a critical regulator for hematopoietic stem cells and erythroid/megakaryocyte development
Magnusson M, et al.
Blood, 109(9), 3687-3696 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service