Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0656

Sigma-Aldrich

HCC-1 human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Hemofiltrate CC chemokine-1, M-CIF, Macrophage Colony Inhibition Factor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in E. coli

assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

1.5-15 EC50 (g/mL)

mol wt

8.7 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CCL14(6358)

Biochem/physiol Actions

HCC-1, a member of the C-C, or β chemokine class, is a 8.7 kDa, non-glycosylated polypeptide of 74 amino acids. Mature HCC-1 is produced by cleavage of a 19 amino acid signal peptide from the precursor HCC-1 (93 amino acids).
HCC-1, a member of the C-C, or β chemokine class, is a 8.7 kDa, non-glycosylated polypeptide of 74 amino acids. Mature HCC-1 is produced by cleavage of a 19 amino acid signal peptide from the precursor HCC-1 (93 amino acids). HCC-1 is expressed constitutively in normal tissues and is present in high concentrations in human plasma. It shows approximately 46% amino acid identity with MIP-1α and MIP-1β, and 29-37% sequence identity with other C-C chemokines.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to chemoattract cultured human monocytes or BaF/3 hCCR1 transfected cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Schulz-Knappe et al.
European journal of medical research, 1(5), 223-236 (1996-02-22)
The structural determination of circulating human peptides is essential to determine their correct posttranslationally processed form. Human hemofiltrate from patients with end stage renal disease is accessible in large quantities and is used as a source for the preparation of
E C Chew et al.
Anticancer research, 17(5A), 3581-3585 (1997-12-31)
The nuclear matrix is the non-chromatin skeleton of the nucleus. This structure contributes to the shape of the nucleus and regulates various nuclear functions. In this study, nuclear matrix proteins of human normal liver, a liver cancer cell line, HepG2
P Schulz-Knappe et al.
The Journal of experimental medicine, 183(1), 295-299 (1996-01-01)
A novel CC chemokine, HCC-1, was isolated from the hemofiltrate of patients with chronic renal failure. HCC-1 has a relative molecular mass of 8,673 and consists of 74 amino acids including four cysteines linked to disulfide bonds. HCC-1 cDNA was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service