Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0416

Sigma-Aldrich

Anti-HOXB5 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HOMEOBOX 2A, Anti-HOMEOBOX B5, Anti-HOX-2A, Anti-HOX2.1, Anti-HU-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using lysates of HEK-293T cells over-expressing human HOXB5

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXB5(3215)
mouse ... Hoxb5(15413)

General description

Hox genes are evolutionarily conserved transcription factors. Homeobox B5 (HOXB5) gene is mapped to human chromosome 17q21.32. HOX proteins have a highly conserved 60-amino-acid region (the homeobox).

Specificity

Anti-HOXB5 recognizes human HOXB5 (also known as HOX-2A, HU-1 and Hox2.

Immunogen

synthetic peptide corresponding to amino acids 103-121 of human HOXB5 conjugated to KLH. The corresponding sequence is identical in mouse.

Application

Anti-HOXB5 antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

One of the major functions of Hox genes seems to be the formation of the body plan during embryonic development. Alteration in the homeobox gene function or expression is implicated in the development of cancers including the breast, prostate, kidney, colon, skin and brain. Studies have shown that HOXB5 is necessary for normal mouse lung branching morphogenesis, and its abnormal regulation causes specific alterations in airway branching. HOXB5 functional impairment is implicated in Hirschsprung′s disease. It also plays a key role in the development of the enteric nervous system. Elimination of the HOX5 gene suppresses β-catenin expression and downstream pathway, thereby inhibiting non-small cell lung cancer (NSCLC) tumor progression.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Zhang et al.
Oncology research, 26(1), 37-44 (2017-03-25)
Homeobox B5 (HOXB5), a member of the HOX gene family, has been shown to play an important role in tumor progression. However, the expression and functional role of HOXB5 in human non-small cell lung cancer (NSCLC) have not been defined.
MaryAnn Vitoria Volpe et al.
Birth defects research. Part A, Clinical and molecular teratology, 82(8), 571-584 (2008-06-17)
Hox transcription factors modulate signaling pathways controlling organ morphogenesis and maintain cell fate and differentiation in adults. Retinoid signaling, key in regulating Hox expression, is altered in pulmonary hypoplasia. Information on pattern-specific expression of Hox proteins in normal lung development
M T Lewis
Breast cancer research : BCR, 2(3), 158-169 (2001-03-16)
Both normal development and neoplastic progression involve cellular transitions from one physiological state to another. Whereas much is being discovered about signal transduction networks involved in regulating these transitions, little progress has been made in identifying the higher order genetic
Yang Yang et al.
Molecular medicine reports, 16(3), 2773-2778 (2017-07-06)
Osteosarcoma is the most common primary malignant bone cancer in children and adolescents. Unfortunately, treatment failures are common due to metastasis and chemore-sistance, however, the underlying molecular mechanism remains unclear. Accumulating evidence indicated that the homeobox B7 (HOXB7) gene was
Nobumoto Tomioka et al.
Cancer genetics and cytogenetics, 201(1), 6-14 (2010-07-17)
Unlike the case with some other solid tumors, whole genome array screening has not revealed prognostic genetic aberrations in primary gastric cancer. Comparative genomic hybridization (CGH) using bacterial artificial chromosome (BAC) arrays for 56 primary gastric cancers resulted in identification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service