Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0409

Sigma-Aldrich

Hylambatin

>96% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$370.00

$370.00


Check Cart for Availability


Select a Size

Change View
5 MG
$370.00

About This Item

Empirical Formula (Hill Notation):
C63H90N16O19S2
Molecular Weight:
1439.61
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$370.00


Check Cart for Availability

Quality Level

assay

>96% (HPLC)

storage temp.

−20°C

SMILES string

FC(F)(F)C(=O)O.S(CC[C@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H]3N(CCC3)C(=O)[C@@H](NC(=O)[C@H]4N(CCC4)C(=O)[C@H]5N(CCC5)C(=O)[C@@H](N)CC(=O)O)CC(=O)O)CC(=O)O)CCCNC(=N)N)Cc2ccccc2)Cc1ccc(cc1)O)CCSC)C(=O)N)C

InChI key

NQOWOPXDLSRKGJ-KKQUMDDGSA-N

Amino Acid Sequence

Asp-Pro-Pro-Asp-Pro-Asp-Arg-Phe-Tyr-Gly-Met-Met-NH2

Biochem/physiol Actions

Tachykinin that increases plasma glucose and plasma insulin.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service