H0285
HIV-1 nef
>95% (SDS-PAGE), recombinant, expressed in E. coli, Galactosidase-tagged
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
recombinant
expressed in E. coli
assay
>95% (SDS-PAGE)
mol wt
20 kDa (plus 114 kDa β-Gal tag)
UniProt accession no.
Storage temp.
−20°C
Gene Information
human immunodeficiency virus 1 ... nef(156110)
General description
Contains amino acid residues [3-190] of HIV-1 nef antigen.
Biochem/physiol Actions
Positive control for HIV antibodies.
Physical form
Solution in 8 M urea; 20 mM Tris-HCL, ph 8.0; 10 mM β−mercaptoethanol.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Lama et al.
Journal of virology, 74(20), 9396-9402 (2000-09-23)
Human immunodeficiency virus (HIV) Nef downregulates the antigen recognition molecules major histocompatibility complex class I and CD4. Downregulation of surface CD4 by Nef relies on the ability of this viral protein to redirect the endocytic machinery to CD4. However, by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service