Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0167

Sigma-Aldrich

HT1171

≥98% (HPLC)

Synonym(s):

5-(2-Methyl-3-nitrothiophen-2-yl)-1,3,4-oxathiazol-2-one, 5-(5-Methyl-4-nitro-2-thienyl)-1,3,4-oxathiazol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4N2O4S2
CAS Number:
Molecular Weight:
244.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cc1sc(cc1[N+]([O-])=O)C2=NSC(=O)O2

InChI

1S/C7H4N2O4S2/c1-3-4(9(11)12)2-5(14-3)6-8-15-7(10)13-6/h2H,1H3

InChI key

ILBBOKHACYTXRK-UHFFFAOYSA-N

Biochem/physiol Actions

Selective inhibitor of mycobacterial proteosomes; 1000-fold selective for mycobacterial proteosomes over human proteosomes.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service