Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0166

Sigma-Aldrich

Anti-HOXB6 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-HOX-2B, Anti-HOX2.2, Anti-HU-2, Anti-Homeo box 2B, Anti-Homeobox B6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using lysates of HEK-293T cells over expressing human HOXB6

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXB6(3216)
mouse ... Hoxb6(15414)

General description

Hox genes are evolutionarily conserved transcription factors. Homeobox B6 (HOXB6) gene is mapped to human chromosome 17q21.32. HOX proteins have a highly conserved 60-amino-acid region (the homeobox).

Specificity

Anti-HOXB6 (N-terminal) recognizes human HOXB6 (also known as HOX2B, HU-2 and Hox-2).

Immunogen

synthetic peptide corresponding to amino acids 48-60 of human HOXB6 conjugated to KLH. The corresponding sequence in mouse differs by one amino acid.

Application

Anti-HOXB6 (N-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

One of the major functions of Hox genes seems to be the formation of the body plan during embryonic development. Alteration in the homeobox gene function or expression is implicated in the development of cancers including the breast, prostate, kidney, colon, skin and brain. HOXB6 is frequently overexpressed in human acute myeloid leukemia (AML). Findings have indicated that HOXB6 overexpression expanded hematopoietic stem cells (HSCs) and myeloid precursors while impairing erythropoiesis and lymphopoiesis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Giampaolo et al.
Leukemia, 16(7), 1293-1301 (2002-07-03)
Homeobox genes encode transcription factors known to be important morphogenic regulators during embryonic development. An increasing body of work implies a role for homeobox genes in both hematopoiesis and leukemogenesis. In the present study we have analyzed the role of
Yang Yang et al.
Molecular medicine reports, 16(3), 2773-2778 (2017-07-06)
Osteosarcoma is the most common primary malignant bone cancer in children and adolescents. Unfortunately, treatment failures are common due to metastasis and chemore-sistance, however, the underlying molecular mechanism remains unclear. Accumulating evidence indicated that the homeobox B7 (HOXB7) gene was
M T Lewis
Breast cancer research : BCR, 2(3), 158-169 (2001-03-16)
Both normal development and neoplastic progression involve cellular transitions from one physiological state to another. Whereas much is being discovered about signal transduction networks involved in regulating these transitions, little progress has been made in identifying the higher order genetic
Neal A Fischbach et al.
Blood, 105(4), 1456-1466 (2004-11-04)
The HOX family of homeobox genes plays an important role in normal and malignant hematopoiesis. Dysregulated HOX gene expression profoundly effects the proliferation and differentiation of hematopoietic stem cells (HSCs) and committed progenitors, and aberrant activation of HOX genes is
Nobumoto Tomioka et al.
Cancer genetics and cytogenetics, 201(1), 6-14 (2010-07-17)
Unlike the case with some other solid tumors, whole genome array screening has not revealed prognostic genetic aberrations in primary gastric cancer. Comparative genomic hybridization (CGH) using bacterial artificial chromosome (BAC) arrays for 56 primary gastric cancers resulted in identification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service