Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22855

Sigma-Aldrich

Anti-Amphiregulin antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Amphiregulin preproprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22855

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AREG(374)

Immunogen

Immunogen Sequence: GI # 4502199, sequence 100-184
Recombinant amphiregulin preproprotein

Application

Anti-Amphiregulin antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Amphiregulin (AREG) is a protein encoded by the AREG gene in humans and is localized to chromosomal region 4q13-4q21, a common breakpoint for acute lymphoblastic leukemia. It is a bifunctional growth modulator that interacts with the EGF/TGF-α receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. The gene is related to epidermal growth factor (EGF) and transforming growth factor α (TGF-α). AREG is a widely expressed transmembrane tyrosine kinase and is synthesized as a membrane-anchored precursor protein that can engage in juxtacrine signaling on adjacent cells. It plays a critical role in the development and maturation of mammary glands, bone tissue and oocytes. Increased expression of the gene is associated with different pathological conditions (mostly of inflammatory and/or neoplastic nature). AREG is useful in primary colorectal tumor as an indicator of prognosis and as a predictor of recurrence.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G D Plowman et al.
Molecular and cellular biology, 10(5), 1969-1981 (1990-05-01)
We have isolated the gene for a novel growth regulator, amphiregulin (AR), that is evolutionarily related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). AR is a bifunctional growth modulator: it interacts with the EGF/TGF-alpha receptor to
Takafumi Ohchi et al.
Anticancer research, 32(6), 2315-2321 (2012-05-30)
Amphiregulin is an epidermal growth factor (EGF) which is a ligand of epidermal growth factor receptor (EGFR). Amphiregulin is the most enhanced EGFR ligand in colon cancer. Here we report on the expression of Amphiregulin using immunohistochemical staining in primary
Carmen Berasain et al.
Seminars in cell & developmental biology, 28, 31-41 (2014-01-28)
Amphiregulin (AREG) is a ligand of the epidermal growth factor receptor (EGFR), a widely expressed transmembrane tyrosine kinase. AREG is synthesized as a membrane-anchored precursor protein that can engage in juxtacrine signaling on adjacent cells. Alternatively, after proteolytic processing by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service