Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22843

Sigma-Aldrich

Anti-TXLNA antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Taxilin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22843

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TXLNA(200081)

Immunogen

Immunogen Sequence: GI # 28460688, sequence 409-539
Recombinant taxilin

Biochem/physiol Actions

α-taxilin, also known as interleukin-14 (IL-14) or high molecular weight B-cell growth factor (HMW-BCGF), is a protein encoded by the TXLNA gene in humans. It belongs to taxilin family that mutually interacts with the syntaxin family members. It acts as a novel binding partner of the syntaxin family, which is implicated in intracellular vesicle traffic. α-taxilin regulate vesicular trafficking and plays an essential role for the release of hepatitis B virus-DNA (HBV-DNA) containing particles. It contains intracellular components that are associated with the microtubule cytoskeleton and functions as a linker protein between the α-taxilin-containing intracellular components and the microtubule cytoskeleton. α-taxilin may also be involved in cell proliferation of hepatocellular carcinomas (HCC) and its expression can be a marker of malignant potential of HCC.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasmin Hoffmann et al.
Journal of hepatology, 59(5), 934-941 (2013-07-03)
α-taxilin was identified as binding partner of syntaxins and is supposed to regulate vesicular trafficking. However, the physiological functions of α-taxilin and its potential relevance for the life cycle of hepatitis B virus (HBV) are still poorly understood. Transfected hepatoma
Satoru Nogami et al.
Biochemical and biophysical research communications, 311(4), 797-802 (2003-11-19)
Taxilin is novel binding partner of the syntaxin family, which is implicated in intracellular vesicle traffic. However, precise binding properties of taxilin to the syntaxin family remain to be clarified. Then, we here further investigated the interaction of taxilin with
Natsuko Ohtomo et al.
International journal of oncology, 37(6), 1417-1423 (2010-11-03)
The membrane traffic system has been recognized to be involved in carcinogenesis and tumor progression in several types of tumors. α-taxilin is a newly identified membrane traffic-related molecule, and its up-regulation has been reported in embryonic and malignant tissues of
Satoru Nogami et al.
Biochemical and biophysical research communications, 319(3), 936-943 (2004-06-09)
The syntaxin family is implicated in intracellular vesicle traffic. We have recently identified taxilin, a novel syntaxin-binding protein, which has a long coiled-coil region in its C-terminal half. A database search has revealed the presence of two other molecules having
Yukimi Horii et al.
Cell structure and function, 37(2), 111-126 (2012-07-13)
Intracellular vesicle traffic plays an essential role in the establishment and maintenance of organelle identity and biosynthetic transport. We have identified α-taxilin as a binding partner of the syntaxin family, which is involved in intracellular vesicle traffic. Recently, we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service