Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22831

Sigma-Aldrich

Anti-ADAMTS-12 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

manufacturer/tradename

Genway 15-288-22831

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

A disintegrin and metalloproteinase with thrombospondin motifs 12 is an enzyme encoded by the ADAMTS12 gene in humans. It belongs to the family of metalloproteinases that helps in mediating a communication between specific cell types and plays a key role in the regulation of normal tissue development, remodeling and degradation.

Immunogen

Immunogen Sequence: GI # 13569928, sequence 297-446
Recombinant A disintegrin-like and metalloprotease with thrombospondin type 1 motif, 12

Application

Anti-ADAMTS-12 antibody produced in chicken is suitbale for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

ADAMTS12 is a degradative enzyme that interacts with the degradable fragments of cartilage oligomeric matrix protein, which is a prominent non-collagenous matrix component in articular cartilage. The protein exhibits a propeptide and metalloproteinase-like, disintegrin-like cysteine-rich domains. It may play a crutial role in pulmonary cells during fetal development or in tumor processes through its proteolytic activity or as a molecule potentially involved in regulation of cell adhesion. ADAMTS12 is found in cartilage, synovial fluid and serum of arthritic patients and may play an important role in the pathogenesis of arthritis. It may be a susceptibile gene for rheumatoid arthritis development. The genetic variations in ADAMTS12 is found to influence the risk of schizophrenia.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina N Bespalova et al.
Neuromolecular medicine, 14(1), 53-64 (2012-02-11)
ADAMTS12 belongs to the family of metalloproteinases that mediate a communication between specific cell types and play a key role in the regulation of normal tissue development, remodeling, and degradation. Members of this family have been implicated in neurodegenerative and
Seong-Su Nah et al.
Molecular medicine reports, 6(1), 227-231 (2012-04-17)
a disintegrin and metalloproteinase (ADAM) with thrombospondin type 1 motif 12 (ADAMTS12) is a degradative enzyme that interacts with the degradable fragments of cartilage oligomeric matrix protein, which is a prominent non-collagenous matrix component in articular cartilage. ADAMTS12 has been
S Cal et al.
The Journal of biological chemistry, 276(21), 17932-17940 (2001-03-30)
We have identified and cloned a human fetal lung cDNA encoding a new protein of the ADAM-TS family (a disintegrin and metalloproteinase domain, with thrombospondin type-1 modules) that has been called ADAM-TS12. This protein exhibits a domain organization similar to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service