Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22828

Sigma-Aldrich

Anti-PDIA6 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat

manufacturer/tradename

Genway 15-288-22828

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDIA6(10130)

General description

PDIA6 (also known as ERP5) is an endoplasmic reticulum (ER)-resident protein found mainly on platelet intracellular membranes. It is translocated to the surface of epithelial tumor cells. It possesses structural homology with protein disulfide isomerase (PDI). PDIA6 consists of two Cys-Gly-His-Cys (CGHC) sequences and the ER retention signal Lys-Asp-Glu-Leu.

Immunogen

Immunogen Sequence: GI # 5031973, sequence 301-440
Recombinant protein disulfide isomerase-associated 6

Application

Anti-PDIA6 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

PDIA6 (also known as ERP5) is involved in various activities such as chaperone, isomerase and peptide-binding activities. It is rapidly recruited to the cell surface in response to a range of platelet agonists. Blocking of cell-surface ERP5 by inhibitory antibodies leads to a decrease in platelet aggregation in response to agonists and a decrease in fibrinogen binding and P-selectin exposure. ERP5 is associated with integrin β(3) subunit during platelet stimulation.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masakazu Kikuchi et al.
Journal of biochemistry, 132(3), 451-455 (2002-09-03)
Human P5 (hP5) was expressed in the Escherichia coli pET system and purified by sequential Ni(2+)-chelating resin column chromatography. Characterization of purified hP5 indicated that it has both isomerase and chaperone activities, but both activities are lower than those of
Leticia Huergo-Zapico et al.
Cancer immunology, immunotherapy : CII, 61(8), 1201-1210 (2012-01-05)
MICA is a ligand of the activating receptor NKG2D, expressed by NK and T cells. MICA expression is induced in cancer cells favoring their elimination by the immune system; however, many advanced tumors shed soluble MICA (sMICA), which impairs NKG2D-mediated
Peter A Jordan et al.
Blood, 105(4), 1500-1507 (2004-10-07)
Formation and rearrangement of disulfide bonds during the correct folding of nascent proteins is modulated by a family of enzymes known as thiol isomerases, which include protein disulfide isomerase (PDI), endoplasmic reticulum protein 5 (ERP5), and ERP57. Recent evidence supports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service