Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22827

Sigma-Aldrich

Anti-ATP5B antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-22827

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP5B(506)

Immunogen

Immunogen Sequence: GI # 4502295, sequence 291-539
Recombinant ATP synthase, H+ transporting, mitochondrial F1 complex, β polypeptide

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirokazu Ishii et al.
Development, growth & differentiation, 56(2), 175-188 (2014-01-15)
In ascidian eggs, cytoplasmic and cortical reorganization, previously called ooplasmic segregation, occurs in two phases during the first cell cycle. In the second phase of reorganization, the mitochondria-rich cytoplasm (myoplasm) moves to the future posterior side, concurrent with sperm aster
Hirokazu Ishii et al.
Development, growth & differentiation, 54(8), 753-766 (2012-10-17)
Previously, we revealed that p58, one of the ascidian maternal factors, is identical to the alpha-subunit of F1-ATP synthase (ATPα), a protein complex of the inner mitochondrial membrane. In the current study, we used immunological probes for ascidian mitochondria components

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service