Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22787

Sigma-Aldrich

Anti-TLR2 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CD282

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-22787

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLR2(7097)

Immunogen

Immunogen Sequence: GI # 19718734, sequence 215-410
Recombinant toll-like receptor 2

Application

Anti-TLR2 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Toll-like receptor 2 is a protein encoded by the TLR2 gene in humans. It is also referred to as CD282 (cluster of differentiation 282). TLR2 is associated with the pathogenesis of inflammatory and noninflammatory acne lesions. It plays a crutial role in innate immune responses and possesses the ability to recognize as well as sense a wide repertoire of infectious and endogenous cellular structural elements. TLR2 also has a potential role in the pathogen-mediated susceptibility to bipolar disorder (BD).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Oliveira et al.
Journal of affective disorders, 165, 135-141 (2014-06-03)
Toll-like receptor 2 (TLR2) molecules play a pivotal role in innate immune responses by their ability to recognize and sense a wide repertoire of infectious and endogenous cellular structural elements. Here we evaluated whether genetic variants in TLR2 influence the
Ola Ahmed Bakry et al.
Analytical and quantitative cytopathology and histopathology, 36(2), 100-110 (2014-06-07)
To elucidate the role of toll-like receptor 2 (TLR2) in the pathogenesis of acne vulgaris through its immunohistochemical localization in inflammatory and noninflammatory lesions of this disease entity. Using standard immunohistochemical techniques, we examined 30 acne cases (involved and noninvolved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service