Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22771

Sigma-Aldrich

Anti-PYY (ab1) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PYY1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-22771

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PYY(5697)
mouse ... Pyy(217212)

Immunogen

Immunogen Sequence: GI # 21703868, sequence 29-40
Synthetic peptide sequence of YY (Mus musculars).

Application

Anti-PYY (ab1) antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Peptide YY (PYY, also known as peptide tyrosine tyrosine) is a peptide encoded by the PYY gene in humans. It is helpful in mediating the beneficial effects of bariatric surgery and plays a role in glucose homeostasis. PYY is affected in various gastrointestinal diseases and disorders.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therese E Fazio Coles et al.
Cell and tissue research, 379(1), 109-120 (2019-09-04)
Recent studies reveal substantial species and regional differences in enteroendocrine cell (EEC) populations, including differences in patterns of hormone coexpression, which limit extrapolation between animal models and human. In this study, jejunal samples, with no histologically identifiable pathology, from patients
Hyun-Jung Cho et al.
Cell and tissue research, 357(1), 63-69 (2014-05-21)
A sub-group of enteroendocrine cells (L cells) release gastrointestinal hormones, GLP-1 and PYY, which have different but overlapping physiological effects, in response to intraluminal nutrients. Whilst their release profiles are not identical, how the plasma levels of these two hormones
Josiane Fakhry et al.
Cell and tissue research, 376(1), 37-49 (2018-11-24)
Gastric endocrine cell hormones contribute to the control of the stomach and to signalling to the brain. In other gut regions, enteroendocrine cells (EECs) exhibit extensive patterns of colocalisation of hormones. In the current study, we characterise EECs in the
Josiane Fakhry et al.
Cell and tissue research, 369(2), 245-253 (2017-04-18)
There is general consensus that enteroendocrine cells, EEC, containing the enteric hormone cholecystokinin (CCK) are confined to the small intestine and predominate in the duodenum and jejunum. Contrary to this, EEC that express the gene for CCK have been isolated
Magdy El-Salhy et al.
International journal of molecular medicine, 31(2), 275-282 (2013-01-08)
Peptide YY (PYY) is affected in several gastrointestinal diseases and disorders. Changes in PYY appear to be an adaptive response to alterations in pathophysiological conditions caused by the disease. This applies to gastrointestinal diseases/disorders such as irritable bowel syndrome, inflammatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service