Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22609

Sigma-Aldrich

Anti-CDKN1B antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

chicken

manufacturer/tradename

Genway 15-288-22609

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... CDKN1B(374106)

Immunogen

Immunogen Sequence: GI # 45383622, sequence 1-198
Recombinant cyclin-dependent kinase inhibitor 1B (p27, Kip1) [Gallus gallus]

Application

Anti-CDKN1B antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Cyclin-dependent kinase inhibitor 1B (p27Kip1) is an enzyme inhibitor that, in humans, is encoded by the CDKN1B gene and belongs to the Cip/Kip family of cyclin dependent kinase (Cdk) inhibitor proteins. It acts as a multifunctional protein to control critical steps of neuronal migration through activities that go far beyond cell cycle regulation. Mutation in CDKN1B gene causes an autosomal dominant disorder called multiple endocrine neoplasia type 4 (MEN4). The cell cycle inhibitor CDKN1B is involved in ERK1/2-mediated MMP-9 expression through activation of the NF-κB binding motif, which leads to the migration and invasion of bladder cancer cells induced by IL-7.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung Lyea Park et al.
International journal of oncology, 44(4), 1349-1356 (2014-02-08)
Interleukin-7 (IL-7) plays a pivotal role in the development and survival of lymphocytes, but its role in cancer cell responses remains unexplained. In this study, IL-7 treatment resulted in a significant induction in the wound-healing migration and Matrigel invasion of
Juliette D Godin et al.
Advances in experimental medicine and biology, 800, 59-74 (2013-11-19)
The cerebral cortex is one of the most intricate regions of the brain, which required elaborated cell migration patterns for its development. Experimental observations show that projection neurons migrate radially within the cortical wall, whereas interneurons migrate along multiple tangential
Francesco Tonelli et al.
European journal of endocrinology, 171(2), K7-K17 (2014-05-14)
Multiple endocrine neoplasia type 4 (MEN4) is an autosomal dominant disorder that presents with a spectrum of clinical manifestations overlapping with those of MEN1 syndrome. It is caused by inactivating mutations of the CDKN1B gene, encoding for p27(kip1) cyclin-dependent kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service