Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22608

Sigma-Aldrich

Anti-WAS antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22608

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WAS(7454)

Immunogen

Immunogen Sequence: GI # 4507909, sequence 313-425
Recombinant wiskott-Aldrich syndrome protein

Application

Anti-WAS antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Wiskott-Aldrich syndrome (WAS) is a rare X-linked recessive disease and is caused by mutations in Wiskott-Aldrich Syndrome Protein (WASP). It is characterized by eczema, thrombocytopenia (low platelet count), immune deficiency, a higher susceptibility to develop tumors and bloody diarrhea (secondary to the thrombocytopenia). WAS is also involved in defects in cell adhesion underpinning cytotoxicity and disease pathogenesis. It regulates the cytoskeleton in hematopoietic cells.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Carmina Castiello et al.
Journal of autoimmunity, 50, 42-50 (2013-12-29)
Wiskott-Aldrich Syndrome protein (WASp) regulates the cytoskeleton in hematopoietic cells and mutations in its gene cause the Wiskott-Aldrich Syndrome (WAS), a primary immunodeficiency with microthrombocytopenia, eczema and a higher susceptibility to develop tumors. Autoimmune manifestations, frequently observed in WAS patients
Shulian Zhang et al.
Fetal and pediatric pathology, 32(4), 312-315 (2013-01-11)
Wiskott-Aldrich syndrome (WAS) is a rare X-linked recessive disease characterized by eczema, thrombocytopenia and immune deficiency. WAS gene mutations impair WAS protein function which cause WAS. The WAS-related disorders of X-linked thrombocytopenia (XLT) and X-linked congenital neutropenia (XLN) may have
Neeraj Jain et al.
Biochimica et biophysica acta, 1842(4), 623-634 (2014-01-21)
Wiskott-Aldrich Syndrome (WAS) is caused by mutations in Wiskott-Aldrich Syndrome Protein (WASP) and majority of the mutations are found in the WASP Homology 1 (WH1) domain which mediates interaction with WIP (WASP Interacting Protein), a WASP chaperone. Two point mutations
Anne-Caroline Chany et al.
Journal of medicinal chemistry, 57(17), 7382-7395 (2014-08-27)
Mycolactone is a complex macrolide toxin produced by Mycobacterium ulcerans, the causative agent of skin lesions called Buruli ulcers. Mycolactone-mediated activation of neural (N) Wiskott-Aldrich syndrome proteins (WASP) induces defects in cell adhesion underpinning cytotoxicity and disease pathogenesis. We describe
Sudha Kumari et al.
The EMBO journal, 39(5), e102783-e102783 (2020-01-03)
When migratory T cells encounter antigen-presenting cells (APCs), they arrest and form radially symmetric, stable intercellular junctions termed immunological synapses which facilitate exchange of crucial biochemical information and are critical for T-cell immunity. While the cellular processes underlying synapse formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service