Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22501

Sigma-Aldrich

Anti-Interleukin-18 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22501

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL18(3606)

Immunogen

Immunogen Sequence: GI # 4504653, sequence 20-155
Recombinant interleukin 18 proprotein

Application

Anti-Interleukin-18 antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Interleukin-18 (IL18, also known as interferon-γ inducing factor) is a protein encoded by the IL18 gene in humans and is mapped to chromosome 11q22.2-q22.3. It is a well-known major proinflammatory cytokine with broad biological effects. Major function of IL-18 involves the enhancement of T cell and natural killer cell cytotoxicity. It is an inflammatory cytokine and plays an important role in autoimmune disease by inducing interferon-γ secretion. IL18 may contribute to SLE (systemic lupus erythematosus) activity.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nehad A Fouad et al.
The Egyptian journal of immunology, 21(1), 1-12 (2014-09-11)
Systemic lupus erythematosus (SLE) is a multisystem autoimmune connective tissue disorder characterized by loss of self-tolerance causing immune-mediated tissue destruction and various clinical presentations Interleukin-18 (IL-18) is a proinflammatory cytokine that plays an important role in chronic inflammation and autoimmune
Mohamed Mandour et al.
Revista da Sociedade Brasileira de Medicina Tropical, 47(2), 137-142 (2014-05-28)
Interleukin (IL)-18 is a well-known major proinflammatory cytokine with broad biological effects. The major immunomodulatory functions of IL-18 include enhancing T cell and natural killer cell cytotoxicity. Serum levels of this cytokine were shown to increase in chronic hepatitis C
The human interleukin 18 gene IL18 maps to 11q22.2-q22.3, closely linked to the DRD2 gene locus and distinct from mapped IDDM loci.
K F Nolan et al.
Genomics, 51(1), 161-163 (1998-08-07)
Huiyuan Li et al.
DNA and cell biology, 33(8), 537-542 (2014-04-20)
Interleukin-18 (IL-18) is an inflammatory cytokine that plays an important role in autoimmune disease by inducing interferon-γ secretion. Considering the abnormal serum concentration of IL-18 in primary immune thrombocytopenia (ITP) patients and the regulated effect of IL-18 gene polymorphisms on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service