Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22431A

Sigma-Aldrich

Anti-NUP62 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22431A

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUP62(23636)

General description

NUP62 (nucleoporin 62kDa) is a component of the nuclear pore complex (NPC), and is located in the central channel. It is a glycosylated FG-Nup (phenylalanine and glycine -rich nucleoporin). It has an NTF2 (nuclear transport factor 2)-docking site at its FG-rich N-terminal region. A Ser/Thr rich linker connects the FG-rich region to the C-terminal, which supposedly is the glycosylation site. The C-terminal acts as an anchor to NPC, and has a coiled-coil conformation.

Immunogen

Immunogen Sequence: GI # 7705355, sequence 328-522
Recombinant nucleoporin 62kDa; nucleoporin p62; nucleoporin 62kD

Application

Anti-NUP62 antibody produced in chicken is suitbale for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Nucleoporin p62 (p62) is synthesized as a soluble cytoplasmic precursor of 61kDa. Nup62 proteins are degraded in infected cells of human rhinovirus (HRV) and poliovirus (PV) and that contributes to the disruption of certain nuclear transport pathways. It is found to be cleaved at multiple sites during viral infection. Loss of this gene leads to aberrant centrosome segregation, and maturation of centriole during the G2 phase. Inactivation of this gene also results in multi-nucleated cells, multi-polar centrosomes, abnormalities in centrosome synthesis and aberrations in spindle orientation. Thus, Nup62 might be involved in the targeting of SAS (spindle assembly)-6 and γ-tubulin to centrioles.[1]

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chieko Hashizume et al.
Cell cycle (Georgetown, Tex.), 12(24), 3804-3816 (2013-10-11)
Centrosomes are comprised of 2 orthogonally arranged centrioles surrounded by the pericentriolar material (PCM), which serves as the main microtubule organizing center of the animal cell. More importantly, centrosomes also control spindle polarity and orientation during mitosis. Recently, we and
Nogi Park et al.
The Journal of biological chemistry, 285(37), 28796-28805 (2010-07-14)
Previous work has shown that several nucleoporins, including Nup62 are degraded in cells infected with human rhinovirus (HRV) and poliovirus (PV) and that this contributes to the disruption of certain nuclear transport pathways. In this study, the mechanisms underlying proteolysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service